Learn More
Gibco™ PBS (10X), pH 7.2
Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
$139.00 - $794.00
Spécifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
|---|---|
| Concentration | 10 X |
| For Use With (Application) | Mammalian Cell Culture, Chromatin Biology |
| pH | 7.1 to 7.3 |
| Physical Form | Liquid |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
70013032
|
Gibco™
70013032 |
500 mL |
chaque for $139.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
70013073
|
Gibco™
70013073 |
10 x 500 mL |
chaque for $794.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ PBS may require pH adjustment at the time of dilution (see protocol for details).
We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.
This PBS is manufactured as follows:
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature
Spécifications
| Phosphate Buffered Saline (PBS) | |
| Mammalian Cell Culture, Chromatin Biology | |
| Liquid | |
| Phosphate Buffered Saline | |
| No Sodium Pyruvate | |
| 2800 - 3200 mOsm/kg |
| 10 X | |
| 7.1 to 7.3 | |
| Gibco™ | |
| Sterile-filtered | |
| cGMP-compliant under the ISO 13485 standard |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.