missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ One Shot™ Fetal Bovine Serum, Qualified, One Shot format, USDA-approved Regions DFS Item

Sera for specialty research and specific assays

$103.00 - $103.00

Specifications

Format One Shot
Grade Qualified
Endotoxin Concentration ≤ 50 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 25 mg/dL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10437077 Life Technologies
LS10437077
50mL Each for $103.00
Specifications

Description

  • Sera Category: Standard
  • Sera for cell culture with robust cell lines
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL
  • Hemoglobin level: ≤25mg/dL
  • Origin: Countries meeting USDA importation requirements
  • ISO 13485 certified, processed in FDA registered facilities
  • Labeled for in-vitro diagnostics (compliant with highest level of USP sterility testing)
  • Triple filtered at 0.1μm
  • Gibco™ bottle is easier to use in hood, reduces risk of contamination and helps to perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Cell Culture, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Sf9 and Sf21 Cell Culture

Order Info

Shipping Condition: Dry Ice

Description & Specifications

Specifications

One Shot
≤ 50 EU/mL
≤ 25 mg/dL
Qualified
No