Please login to your online account to display your discounted pricing

Gibco™ MIP-1-β Recombinant Human Protein DFS Item

Recombinant MIP-1-beta is a bioactive protein intended for use in cell culture applications

$247.80 - $247.80

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Liquid
Gene Alias MIP-1β
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHC1034Z

DFS Item
 
gibco™
PHC1034Z
10μg Each for $247.80
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Liquid
Gene Alias MIP-1β
Gene ID (Entrez) 6351

MIP-1-beta (Macrophage inflammatory protein-1β) is involved in a number of biological processes including chemotaxis and is involved in inducing the synthesis of pro-inflammatory cytokines including TNF, IL-6, and IL-1.

Shipping Condition: Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.