Please login to your online account to display your discounted pricing

Gibco™ MIP-1-β Recombinant Human Protein DFS Item


Recombinant MIP-1-beta is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC1034Z

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC1034Z

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 8kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 10μg
Source Human
Storage Requirements Store at -5° to -30°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype MIP (Macrophage Inflammatory Protein) Chemokines
Endotoxin Concentration <0.1ng/μg
Gene Alias MIP-1β
Validated Application Cell Culture
Protein Family Chemokines & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture; Drug Discovery & Development; Target & Lead Identification & Validation; Kinase Biology; Biochemical Kinase Assays
Format Liquid
Gene ID (Entrez) 6351

MIP-1-beta (Macrophage inflammatory protein-1β) is involved in a number of biological processes including chemotaxis and is involved in inducing the synthesis of pro-inflammatory cytokines including TNF, IL-6, and IL-1.

Shipping Condition: Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.