missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ MEM, No Glutamine DFS Item

Minimum Essential Medium is commonly used of all cell culture media

$41.40 - $356.00

Specifications

Format Liquid
Formulation Calcium Chloride (CaCl2) (anhyd.), Choline chloride, D-Calcium pantothenate, Folic Acid, L-Arginine hydrochloride, L-Cystine 2HCl, L-Histidine hydrochloride-H2O, L-Isoleucine, L-Leucine, L-Lysine hydrochloride, L-Methionine, L-Phenylalanine, L-Threonine, L-Tryptophan, L-Tyrosine disodium salt dihydrate, L-Valine, Magnesium Sulfate (MgSO4) (anhyd.), Niacinamide, Potassium Chloride (KCl), Pyridoxal hydrochloride, Riboflavin, Sodium Bicarbonate (NaHCO3), Sodium Chloride (NaCl), Sodium Phosphate monobasic (NaH2PO4-H2O), Thiamine hydrochloride, i-Inositol
Packaging Sustainable packaging
Storage Requirements Storage: 2-8° C. Protect from light. Shelf life: 12 months from date of manufacture
View More Specs

Disclaimers:

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11090099 Life Technologies
LS11090099
10 x 500mL Each for $356.00
11090073 Life Technologies
LS11090073
1000mL Each for $70.50
11090081 Life Technologies
LS11090081
500mL Each for $41.40
Specifications

Description

MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. Gibco MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification.

  • Modified as follows: with phenol red; without L-glutamine, HEPES
  • Available with Earle's salts for use in CO2 incubator, or with Hank's salts for use without CO2
  • Made with Earle's salts
  • Contains no proteins, lipids, or growth factors
  • Requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • Uses sodium bicarbonate buffer system (2.2g/L) and therefore requires 5-10% CO2 environment to maintain physiological pH
  • Sustainable packaging

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Liquid
Calcium Chloride (CaCl2) (anhyd.), Choline chloride, D-Calcium pantothenate, Folic Acid, L-Arginine hydrochloride, L-Cystine 2HCl, L-Histidine hydrochloride-H2O, L-Isoleucine, L-Leucine, L-Lysine hydrochloride, L-Methionine, L-Phenylalanine, L-Threonine, L-Tryptophan, L-Tyrosine disodium salt dihydrate, L-Valine, Magnesium Sulfate (MgSO4) (anhyd.), Niacinamide, Potassium Chloride (KCl), Pyridoxal hydrochloride, Riboflavin, Sodium Bicarbonate (NaHCO3), Sodium Chloride (NaCl), Sodium Phosphate monobasic (NaH2PO4-H2O), Thiamine hydrochloride, i-Inositol
Sustainable packaging
Storage: 2-8° C. Protect from light. Shelf life: 12 months from date of manufacture