missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ MCP-1 (MCAF) Recombinant Rat Protein DFS Item

Recombinant MCP-1 (MCAF) is a bioactive protein intended for use in cell culture applications

$233.00 - $1,048.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias MCP-1, MCAF
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRC1011 Gibco™
LSPRC1011
100μg Each for $1,048.00
PRC1014 Gibco™
LSPRC1014
10μg Each for $233.00
PRC1015 Gibco™
LSPRC1015
25μg Each for $446.00
Specifications

Description

MCP-1 is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

95%
<0.1ng/μg
MCP-1, MCAF
Sequential Chromatography
Lyophilized
24770