Please login to your online account to display your discounted pricing

Gibco™ MCP-1 (MCAF) Recombinant Rat Protein DFS Item


Recombinant MCP-1 (MCAF) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPRC1015

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PRC1015

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 14kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 25μg
Source Rat
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype MCP/MCAF Chemokines
Endotoxin Concentration <0.1ng/μg
Gene Alias MCP-1, MCAF
Validated Application Cell Culture
Protein Family Chemokines & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture
Format Lyophilized
Gene ID (Entrez) 24770

MCP-1 is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.