Please login to your online account to display your discounted pricing

Gibco™ MCP-1 (MCAF) Recombinant Rat Protein DFS Item


Recombinant MCP-1 (MCAF) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPRC1011

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PRC1011

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at 2° to 8°C.
For Use With (Application) Cell Culture; Mammalian Cell Culture
Quantity 100μg
Source Rat
Molecular Weight 14kDa
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Family Chemokines & Receptors
Purity 95%
Protein Subtype MCP/MCAF Chemokines
Validated Application Cell Culture
Gene Alias MCP-1, MCAF
Endotoxin Concentration <0.1ng/μg
Expression System E. coli
Purification Method Sequential Chromatography
Gene ID (Entrez) 24770
Format Lyophilized

MCP-1 is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.