Please login to your online account to display your discounted pricing

Gibco™ M-CSF Recombinant Human Protein DFS Item

M-CSF (macrophage colony-stimulating factor) is a bioactive protein intended for use in cell culture applications

$317.00 - $1,676.00

Specifications

Purity >95% by SDS-PAGE
Endotoxin Concentration <0.1ng/μg
Gene Alias M-CSF
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHC9501

DFS Item
 
gibco™
LSPHC9501
100μg Each for $1,676.00

PHC9504

DFS Item
 
gibco™
LSPHC9504
10μg Each for $317.00
Description & Specifications

Specifications

Purity >95% by SDS-PAGE
Endotoxin Concentration <0.1ng/μg
Gene Alias M-CSF

M-CSF induces proliferation and differentiation of hematopoietic stem cells into macrophages and growth and differentiation of monocytes.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: proteins bioassayed with Gibco media
  • High-purity, recombinant, bioactive protein: to help ensure our growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High biological activity: in-house activity testing of our recombinant proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the dose dependent stimulation of proliferation of mouse M-NFS-60 cells

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.