missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Human LIF Recombinant Protein
Recombinant human leukemia inhibitory factor (LIF) is a bioactive protein intended for use in cell culture applications
Supplier: Gibco™ PHC9484
Description
Human LIF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
LIF is a cytokine that induces macrophage differentiation. LIF, in conjunction with BMP2, acts on primary fetal neural progenitor cells to induce astrocytes.- High purity: minimizes interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco recombinant proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the LIF dose-dependent proliferation of Human TF-1 cells
Shipping Condition: Dry Ice.
Specifications
21 kDa | |
10μg | |
LIF | |
Untagged | |
0.5 ng/ml | |
Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Oncology | |
95 % |
Sequential Chromatography | |
LIF | |
Human | |
Lyophilized form should be stored at 2–8°C, preferably desiccated. Reconstituted form should be stored at ≤ –20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum. |
|
Dose-dependent proliferation of TF-1 cells | |
E. coli | |
Recombinant, Ligand | |
Other Cytokines | |
Gibco™ | |
Leukemia inhibitory factor |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.