Please login to your online account to display your discounted pricing

Gibco™ Kallikrein 7 Recombinant Human Protein DFS Item

Recombinant Kallikrein 7 is a bioactive protein intended for use in cell culture applications

$372.00 - $2,080.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias Kallikrein 7
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHC9291

DFS Item
 
gibco™
LSPHC9291
100μg Each for $2,080.00

PHC9294

DFS Item
 
gibco™
LSPHC9294
10μg Each for $372.00
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias Kallikrein 7
Gene ID (Entrez) KLK7
Accession Number NM_005046

KLK7 is one of the Kallikreins, which are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.