missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Kallikrein 7 Recombinant Human Protein DFS Item

Recombinant Kallikrein 7 is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPHC9291

 View more versions of this product

Catalog No. PHC9291


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Gibco; Kallikrein 7 Recombinant Human Protein; 100ug; Source: Human; Purity: 95%

KLK7 is one of the Kallikreins, which are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis.

Shipping Condition: Wet Ice.

Description & Specifications

Specifications

NM_005046
Lyophilized
30kDa
95%
Human
Carrier Protein
TGFs (Transforming Growth Factors)
Kallikrein 7
Growth Factors & Receptors
300pmole/min./μg
Untagged
Cell Culture; Mammalian Cell Culture
KLK7
Sequential Chromatography
100μg
Lyophilized Human Kallikrein 7 should be stored at 2° to 8°C, preferably desiccated. Store reconstituted Human Kallikrein 7 at ≤ -20°C. Avoid freeze/thaw cycles.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
Mammalian
Human