Please login to your online account to display your discounted pricing

Gibco™ IP10 Recombinant Mouse Protein DFS Item


Recombinant IP10 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC1083

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PMC1083

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 8.7kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 1mg
Source Mouse
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype IP-10
Endotoxin Concentration <0.1ng/μg
Gene Alias IP-10
Validated Application Cell Culture
Protein Family Chemokines & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture
Format Lyophilized

IP10 is a chemokine of the CXC subfamily and ligand for the receptor CXCR3. Binding of this protein to CXCR3 results in pleiotropic effects, including stimulation of monocytes, natural killer and T-cell migration, and modulation of adhesion molecule expression.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.