missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ IP10 Recombinant Human Protein DFS Item

Recombinant IP10 is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPHC1084

 View more versions of this product

Catalog No. PHC1084


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Gibco; IP10 Recombinant Human Protein; 10ug; Source: Human; Purity: 95%

IP10 is a chemokine of the CXC subfamily and ligand for the receptor CXCR3. Binding of this protein to CXCR3 results in pleiotropic effects, including stimulation of monocytes, natural killer and T-cell migration, and modulation of adhesion molecule expression.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Description & Specifications

Specifications

Cell Culture; Mammalian Cell Culture
3627
Sequential Chromatography
10μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
∽8kDa
95%
Human
Carrier Protein
IP-10
IP-10
Chemokines & Receptors