Please login to your online account to display your discounted pricing

Gibco™ IP10 Recombinant Human Protein DFS Item


Recombinant IP10 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC1083

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC1083

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at 2° to 8°C.
Quantity 1mg
Molecular Weight ∽8kDa
Protein Family Chemokines & Receptors
Expression System E. coli
Validated Application Cell Culture
Endotoxin Concentration <0.1ng/μg
Purity 95%
Gene Alias IP-10
Purification Method Sequential Chromatography
Source Human
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype IP-10
For Use With (Application) Cell Culture; Mammalian Cell Culture
Format Lyophilized

IP10 is a chemokine of the CXC subfamily and ligand for the receptor CXCR3. Binding of this protein to CXCR3 results in pleiotropic effects, including stimulation of monocytes, natural killer and T-cell migration, and modulation of adhesion molecule expression.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.