missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ IMDM, GlutaMAX™ Supplement DFS Item

Iscove's Modified Dulbecco's Medium is well suited for rapidly proliferating, high-density cell cultures including Jurkat, COS-7, and macrophage cells

Manufacturer:  Gibco™ 31980030

 View more versions of this product

Catalog No. 31980030



Requires : Supplementation

Description

Description

  • IMDM (Iscove's Modified Dulbecco's Medium) is well suited for rapidly proliferating, high-density cell cultures including Jurkat, COS-7, and macrophage cells
  • Store at 2°C to 8°C, protected from light

Modified as follows:

  • With: GlutaMAX™, phenol red
  • Without: α-thioglycerol, 2-mercaptoethanol

Using IMDM

  • IMDM, a modification of Dulbecco's Modified Eagle Medium, includes selenium as well as additional amino acids and vitamins
  • This unique medium lacks iron, with potassium nitrate replacing ferric nitrate
  • IMDM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
  • IMDM contains no proteins, lipids, or growth factors therefore it requires supplementation, commonly with 10% Fetal Bovine Serum (FBS)
  • It uses a sodium bicarbonate buffer system (3.024 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH
Specifications

Specifications

1X
Sterile-filtered
Room Temperature
Liquid
Phenol Red
Sustainable packaging
GlutaMAX-I, Phenol Red
High Glucose
Jurkat, COS-7, and macrophage cells
IMDM (Iscove's Modified Dulbecco's Medium)
IMDM (Iscove's Modified Dulbecco's Medium)
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Gibco™, GlutaMAX™
GlutaMAX-I
12 Months From Date of Manufacture
500 mL
α-Thioglycerol, 2-Mercaptoethanol
Sodium Pyruvate
Animal Origin-Free
HEPES
SDS
Documents
Product Certifications
Promotions

Promotions

In vitro diagnostic