Please login to your online account to display your discounted pricing

Gibco™ IL9 Recombinant Human Protein DFS Item


Recombinant Human Interleukin 9 (IL9) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC0094

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PHC0094

  • / Each
 

Description & Specifications

Specifications

Purity 95%
Quantity 10μg
Source Human
Storage Requirements Store at -5° to -30°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Endotoxin Concentration <1 EU/mL
Gene Alias IL-9
Assays Proliferation of human MO7e cells
Validated Application Cell Culture
Expression System Mammalian
Biological Activity 0.2ng/mL
Format Lyophilized
Molecular Weight 48kDa
Protein Subtype Interleukins
Protein Family Cytokines & Receptors
For Use With (Application) Cell Culture; Mammalian Cell Culture

IL9 is produced by CD4+ helper cells, and has been shown to be a pleiotropic cytokine impacting lymphoid and mast cell lineages.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to get a biologically active protein
  • High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose-dependent stimulation of the proliferation of human MO7e cells
  • Proven compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.