missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ IL8 Monocyte Recombinant Human Protein DFS Item

Recombinant IL8 Monocyte is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPHC0885

 View more versions of this product

Catalog No. PHC0885



Description

Description

IL8 is chemotactic for all known types of migratory immune cells. IL8 also inhibits the adhesion of leukocytes to activated endothelial cells.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
3576
Sequential Chromatography
50μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
8.6kDa
95%
Human
Carrier Protein
Interleukins
IL-8 Monocyte
Chemokines & Receptors
Documents

For Research Use Only. Not for use in diagnostic procedures.