Please login to your online account to display your discounted pricing

Gibco™ IL8 Monocyte Recombinant Human Protein DFS Item


Recombinant IL8 Monocyte is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC0884

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC0884

  • / Each
 

Description & Specifications

Specifications

Quantity 25μg
Expression System E. coli
Validated Application Cell Culture
Gene Alias IL-8 Monocyte
Protein Family Chemokines & Receptors
Protein Subtype Interleukins
Source Human
Structural Form Recombinant/Ligand
For Use With (Application) Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
Gene ID (Entrez) 3576
Storage Requirements Store at 2° to 8°C.

IL8 is chemotactic for all known types of migratory immune cells. IL8 also inhibits the adhesion of leukocytes to activated endothelial cells.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.