Please login to your online account to display your discounted pricing

Gibco™ IL8 Monocyte Recombinant Human Protein DFS Item


Recombinant IL8 Monocyte is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC0881

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC0881

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at 2° to 8°C.
Protein Family Chemokines & Receptors
Purification Method Sequential Chromatography
Format Lyophilized
Gene ID (Entrez) 3576
Source Human
Expression System E. coli
Protein Subtype Interleukins
Without Additives Carrier Protein
Molecular Weight 8.6kDa
Quantity 100μg
Structural Form Recombinant/Ligand
Endotoxin Concentration <0.1ng/μg
Gene Alias IL-8 Monocyte
Validated Application Cell Culture
For Use With (Application) Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
Purity 95%

IL8 is chemotactic for all known types of migratory immune cells. IL8 also inhibits the adhesion of leukocytes to activated endothelial cells.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.