Please login to your online account to display your discounted pricing

Gibco™ IL6 Recombinant Mouse Protein DFS Item


Recombinant Mouse Interleukin 6 (IL6) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC0061

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PMC0061

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 16193
Protein Subtype Interleukins
Source Mouse
Purity 95%
For Use With (Application) Cell Culture; Mammalian Cell Culture
Endotoxin Concentration <0.1ng/μg
Expression System E. coli
Gene Alias IL-6
Validated Application Cell Culture
Storage Requirements Store at 2° to 8°C.
Molecular Weight ∽20kDa
Protein Family Cytokines & Receptors
Purification Method Sequential Chromatography
Format Lyophilized
Structural Form Recombinant/Ligand
Quantity 100μg
Without Additives Carrier Protein

IL6 is a pleiotropic cytokine involved in several cellular processes including inflammation, hematopoeisis, and immune response.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse B9 cells

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.