Please login to your online account to display your discounted pricing

Gibco™ IL4 Recombinant Swine Protein DFS Item


Recombinant IL4 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPSC0045

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PSC0045

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 12.5kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 25μg
Source Swine/Porcine
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype Interleukins
Endotoxin Concentration <0.1ng/μg
Gene Alias IL-4
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture
Format Lyophilized
Gene ID (Entrez) 397225

IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.