Please login to your online account to display your discounted pricing

Gibco™ IL4 Recombinant Swine Protein DFS Item


Recombinant IL4 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPSC0041

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PSC0041

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 12.5kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 100μg
Source Swine/Porcine
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype Interleukins
Endotoxin Concentration <0.1ng/μg
Gene Alias IL-4
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Angiogenesis; Cell Proliferation
Format Lyophilized

IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors.

Approved for shipment on Wet or Dry Ice.