Please login to your online account to display your discounted pricing

Gibco™ IL4 Recombinant Mouse Protein DFS Item


Recombinant Mouse Interleukin 4 (IL4) is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC0043

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PMC0043

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -5° to -30°C.
Molecular Weight 14kDa
Expression System E. coli
Format Lyophilized
Structural Form Recombinant/Ligand
Protein Subtype Interleukins
Gene ID (Entrez) 10946583, 3565
Purification Method Sequential Chromatography
Source Mouse
Purity 95%
Quantity 1mg
Gene Alias IL-4
Validated Application Cell Culture
Protein Family Cytokines & Receptors
For Use With (Application) Cell Culture; Mammalian Cell Culture

IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the dose-dependent proliferation of murine HT-2 cells

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.