Please login to your online account to display your discounted pricing

Gibco™ IL33 Recombinant Mouse Protein DFS Item

Recombinant IL33 is a bioactive protein intended for use in cell culture applications

$276.00 - $6,950.00

Specifications

Purity 95%
Purification Method PAGE
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-33
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PMC4043

DFS Item
 
gibco™
LSPMC4043
1mg Each for $6,950.00

PMC4044

DFS Item
 
gibco™
LSPMC4044
10μg Each for $276.00
Description & Specifications

Specifications

Purity 95%
Purification Method PAGE
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-33
Gene ID (Entrez) IL33
Accession Number NM_006853

IL33 is a proinflammatory cytokine with structural homology similar with IL-1 family cytokines. IL33 exerts its biological effects by binding to the receptor ST2 and forming a complex with IL-1 Receptor Accessory Protein. Intracellular proteins within in the NFκB and MAP kinase signaling pathways are activated leading to the production of the type 2 cytokines IL-5 from polarized TH2 cells.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.