Please login to your online account to display your discounted pricing

Gibco™ IL33 Recombinant Human Protein DFS Item


Recombinant IL33 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC9251

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC9251

  • / Each
 

Description & Specifications

Specifications

Accession Number NM_033439
Molecular Weight 18kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 100μg
Source Human
Storage Requirements Lyophilized Human IL-33 should be stored at 2° to 8°C, preferably desiccated. Store reconstituted Human IL-33 at ≤ -20°C. Avoid freeze/thaw cycles.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype Interleukins
Endotoxin Concentration <0.1ng/μg
Gene Alias IL-33
Gene Symbol IL33
Assays Proliferation of Mouse D10S Cells
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System E. coli
Protein Length 159 aa
Biological Activity ED50 range: 0.1 to 1.0ng/mL
Specific Reactivity 1.0 x 107 - 1.0 x 106 units/mg
Cross Reactivity Human
For Use With (Application) Cell Culture; Mammalian Cell Culture
Format Lyophilized
Gene ID (Entrez) 90865

IL33 is a proinflammatory cytokine with structural homology similar with IL-1 family cytokines. IL33 exerts its biological effects by binding to the receptor ST2 and forming a complex with IL-1 Receptor Accessory Protein. Intracellular proteins within in the NFκB and MAP kinase signaling pathways are activated leading to the production of the type 2 cytokines IL-5 from polarized TH2 cells.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.