missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Mouse IL-3 Recombinant Protein
Recombinant Mouse Interleukin 3 (IL3) is a bioactive protein intended for use in cell culture applications
Supplier: Gibco™ PMC0033
Description
Mouse IL-3 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL3 provides the cytokine connection between the immune system and the hematopoietic system. For example, IL3 supports the proliferation and development of almost all types of hematopoietic progenitor cells and can act as a chemoattractant for eosinophils.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of NFS-60 cells
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
16.1 kDa | |
1mg | |
Mouse | |
Carrier-Free | |
1 mg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
16187 | |
Sequential Chromatography | |
IL-3 | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.