Please login to your online account to display your discounted pricing

Gibco™ IL20 Recombinant Human Protein DFS Item

Recombinant IL20 is a bioactive protein intended for use in cell culture applications

$269.85 - $269.85

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-20
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHC0204

DFS Item
 
gibco™
PHC0204
10μg Each for $269.85
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-20
Gene ID (Entrez) 50604

There are two heterodimeric receptor complexes for IL20. The first is composed of IL20 Ra and IL20 Rb. The second is composed of IL-22 R and IL20b. IL20 induces the proliferation of multipotential hematopoietic progenitor cells, directs the differentiation and expansion of keratinocytes, and promotes the release of proinflammatory mediators in keratinocytes and other cells expressing the IL20 receptor.

Approved for shipment on Wet or Dry Ice.