Please login to your online account to display your discounted pricing

Gibco™ IL20 Recombinant Human Protein DFS Item


Recombinant IL20 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC0204

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC0204

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at 2° to 8°C.
For Use With (Application) Cell Culture; Mammalian Cell Culture
Gene ID (Entrez) 50604
Molecular Weight ∽18kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 10μg
Source Human
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype Interleukins
Endotoxin Concentration <0.1ng/μg
Gene Alias IL-20
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System E. coli
Format Lyophilized

There are two heterodimeric receptor complexes for IL20. The first is composed of IL20 Ra and IL20 Rb. The second is composed of IL-22 R and IL20b. IL20 induces the proliferation of multipotential hematopoietic progenitor cells, directs the differentiation and expansion of keratinocytes, and promotes the release of proinflammatory mediators in keratinocytes and other cells expressing the IL20 receptor.

Approved for shipment on Wet or Dry Ice.