Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

Gibco™ IL2 Recombinant Mouse Protein DFS Item

Recombinant IL-2 is a bioactive protein intended for use in cell culture applications

$169.00 - $3,420.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-2
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
PMC0021 Gibco™
LSPMC0021
100μg Each for $752.00
PMC0023 Gibco™
LSPMC0023
1mg Each for $3,420.00
PMC0024 Gibco™
LSPMC0024
10μg Each for $169.00
PMC0025 Gibco™
LSPMC0025
25μg Each for $307.00
Specifications

Description

IL-2 causes proliferation of T cells and is a central regulator of immune responses.

Shipping Condition: Wet Ice.

Description & Specifications

Specifications

95%
<0.1ng/μg
IL-2
Sequential Chromatography
Lyophilized
16183