missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ IL2 Recombinant Mouse Protein DFS Item

Recombinant IL-2 is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPMC0025

 View more versions of this product

Catalog No. PMC0025


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Gibco; IL2 Recombinant Mouse Protein; 25ug; Source: Mouse; Purity: 95%

IL-2 causes proliferation of T cells and is a central regulator of immune responses.

Shipping Condition: Wet Ice.

Description & Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
16183
Sequential Chromatography
25μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
∽17kDa
95%
Mouse
Carrier Protein
Interleukins
IL-2
Cytokines & Receptors