Please login to your online account to display your discounted pricing

Gibco™ IL2 Recombinant Mouse Protein DFS Item


Recombinant IL-2 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC0024

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PMC0024

  • / Each
 

Description & Specifications

Specifications

Molecular Weight ∽17kDa
Protein Family Cytokines & Receptors
Gene ID (Entrez) 16183
Validated Application Cell Culture
Gene Alias IL-2
Format Lyophilized
For Use With (Application) Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
Protein Subtype Interleukins
Without Additives Carrier Protein
Purity 95%
Quantity 10μg
Expression System E. coli
Storage Requirements Store at 2° to 8°C.
Purification Method Sequential Chromatography
Source Mouse
Structural Form Recombinant/Ligand
Endotoxin Concentration <0.1ng/μg

IL-2 causes proliferation of T cells and is a central regulator of immune responses.

Shipping Condition: Wet Ice.