Please login to your online account to display your discounted pricing

Gibco™ IL2 Recombinant Mouse Protein DFS Item


Recombinant IL-2 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC0021

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PMC0021

  • / Each
 

Description & Specifications

Specifications

Source Mouse
Purity 95%
Without Additives Carrier Protein
Gene ID (Entrez) 16183
Format Lyophilized
Expression System E. coli
Quantity 100μg
For Use With (Application) Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
Structural Form Recombinant/Ligand
Endotoxin Concentration <0.1ng/μg
Validated Application Cell Culture
Purification Method Sequential Chromatography
Storage Requirements Store at 2° to 8°C.
Gene Alias IL-2
Protein Family Cytokines & Receptors
Molecular Weight ∽17kDa
Protein Subtype Interleukins

IL-2 causes proliferation of T cells and is a central regulator of immune responses.

Shipping Condition: Wet Ice.