Please login to your online account to display your discounted pricing

Gibco™ IL2 Recombinant Human Protein DFS Item

Recombinant Human Interleukin 2 (IL2) is a bioactive protein intended for use in cell culture applications

$231.00 - $1,100.00

Specifications

Purity 95%
Purification Method Membrane-Filtered
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-2
View More Specs

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHC0021

DFS Item
 
gibco™
LSPHC0021
100μg Each for $916.00

PHC0023

DFS Item
 
gibco™
LSPHC0023
1mg Each for $1,100.00

PHC0026

DFS Item
 
gibco™
LSPHC0026
10μg Each for $231.00

PHC0027

DFS Item
 
gibco™
LSPHC0027
40μg Each for $682.50
Description & Specifications

Specifications

Purity 95%
Purification Method Membrane-Filtered
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-2
Gene ID (Entrez) 3558

IL2 causes proliferation of T-cells and is a central regulator of immune responses.

  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse CTLL-2 cells
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.