Please login to your online account to display your discounted pricing

Gibco™ IL1B Recombinant Rat Protein DFS Item


Recombinant IL1B is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPRC0815

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PRC0815

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 17.3kDa
Purity 95%
Source Rat
For Use With (Application) Cell Culture; Mammalian Cell Culture
Purification Method Sequential Chromatography
Structural Form Recombinant/Ligand
Endotoxin Concentration <0.1ng/μg
Storage Requirements Store at 2° to 8°C.
Format Lyophilized
Gene Alias IL-1β
Expression System E. coli
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Protein Subtype Interleukins
Without Additives Carrier Protein
Quantity 25μg
Gene ID (Entrez) 24494

IL-1 is involved in a number of biological process including the stimulation of T-helper cells, promotion proliferation and synthesis of immunoglobulins, and the activation of fibroblasts and NK cells.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.