missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Mouse IL-1 beta Recombinant Protein
Recombinant IL1B is a bioactive protein intended for use in cell culture applications
Supplier: Gibco™ PMC0814
Description
Mouse IL-1 beta Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL-1 is involved in a number of biological process including the stimulation of T-helper cells, promotion proliferation and synthesis of immunoglobulins, and the activation of fibroblasts and NK cells.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
Sequential Chromatography | |
IL-1β | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Stem Cell Research, Angiogenesis | |
95 % |
17 kDa | |
5μg | |
Mouse | |
Carrier-Free | |
5 μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.