Please login to your online account to display your discounted pricing

Gibco™ IL1B Recombinant Mouse Protein DFS Item

Recombinant IL1B is a bioactive protein intended for use in cell culture applications

$302.00 - $2,840.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-1β
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PMC0811

DFS Item
 
gibco™
LSPMC0811
100μg Each for $2,840.00

PMC0814

DFS Item
 
gibco™
LSPMC0814
5μg Each for $302.00

PMC0815

DFS Item
 
gibco™
LSPMC0815
10μg Each for $514.00

PMC0816

DFS Item
 
gibco™
LSPMC0816
25μg Each for $1,056.00
Description & Specifications

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-1β
Gene ID (Entrez) 16176

IL-1 is involved in a number of biological process including the stimulation of T-helper cells, promotion proliferation and synthesis of immunoglobulins, and the activation of fibroblasts and NK cells.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.