missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ IL1A Recombinant Human Protein DFS Item

Recombinant IL1A is a bioactive protein intended for use in cell culture applications

$214.00 - $7,010.00

Specifications

Purity 95%
Purification Method Membrane-Filtered
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-1α
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHC0011 Gibco™
LSPHC0011
100μg Each for $1,568.00
PHC0013 Gibco™
LSPHC0013
1mg Each for $7,010.00
PHC0015 Gibco™
LSPHC0015
5μg Each for $214.00
PHC0017 Gibco™
LSPHC0017
25μg Each for $680.00
Specifications

Description

IL-1 is involved in a number of biological processes including the stimulation of T-helper cells, promotion proliferation and synthesis of immunoglobulins, and the activation of fibroblasts and NK cells.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

95%
<0.1ng/μg
IL-1α
Membrane-Filtered
Lyophilized