missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ IL1A Recombinant Human Protein DFS Item

Recombinant IL1A is a bioactive protein intended for use in cell culture applications

Manufacturer:  Gibco™ LSPHC0017

 View more versions of this product

Catalog No. PHC0017



Description

Description

IL-1 is involved in a number of biological processes including the stimulation of T-helper cells, promotion proliferation and synthesis of immunoglobulins, and the activation of fibroblasts and NK cells.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
3552
Membrane-Filtered
25μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
18.2kDa
95%
Human
Carrier Protein
Interleukins
IL-1α
Cytokines & Receptors
Documents

For Research Use Only. Not for use in diagnostic procedures.