Please login to your online account to display your discounted pricing

Gibco™ IL17F Recombinant Human Protein DFS Item


Recombinant Human IL17F is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC9551

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

Catalog No. PHC9551

  • / Each
 

Description & Specifications

Specifications

Source Human
Molecular Weight 38kDa
Protein Length 133 aa
Expression System Mammalian
Assays Induction of IL-6 secretion from NDHF cells
Validated Application Cell Culture
Biological Activity 100ng/mL
Gene Alias IL-17F
Protein Family Cytokines & Receptors
Endotoxin Concentration <1 EU/mL
Purity 90%
Quantity 100μg
Protein Subtype Interleukins
Purification Method Gel-Purified
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
For Use With (Application) Cell Culture; Mammalian Cell Culture
Storage Requirements Store at -5° to -30°C.

IL17F is a member of structurally related cytokines. The IL-17 family of cytokines plays a role in mediating proinflamatory responses.

  • High purity: minimal interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to help ensure a biologically active protein
  • High biological activity: in house activity testing includes a variety of cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose-dependent induction of IL-6 secretion from NDHF Adult fibroblasts

Shipping Condition: Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.