missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Human IL-17A Recombinant Protein
Recombinant IL17A is a bioactive protein intended for use in cell culture applications
Supplier: Gibco™ PHC9174
Description
Human IL-17A Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL17A is a proinflammatory cytokine that regulates the activities of NFκB and mitogen-activated protein kinases. This cytokine can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO).Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
Sequential Chromatography | |
IL-17A | |
Human | |
Carrier-Free | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
15.5 kDa | |
10μg | |
272 aa (homodimer) | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Neurobiology, Inflammation, Angiogenesis, Signal Transduction | |
95 % |
For Research Use Only. Not for use in diagnostic procedures.