Please login to your online account to display your discounted pricing

Gibco™ IL17 Recombinant Mouse Protein DFS Item


Recombinant IL17 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC0175

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PMC0175

  • / Each
 

Description & Specifications

Specifications

Quantity 25μg
Storage Requirements Store at 2° to 8°C.
For Use With (Application) Cell Culture; Mammalian Cell Culture
Protein Family Cytokines & Receptors
Molecular Weight 15kDa
Structural Form Recombinant/Ligand
Protein Subtype Interleukins
Endotoxin Concentration <0.1ng/μg
Purification Method Sequential Chromatography
Source Mouse
Gene Alias IL-17
Gene ID (Entrez) 16171
Expression System E. coli
Format Lyophilized
Purity 95%
Without Additives Carrier Protein
Validated Application Cell Culture

IL17 stimulates epithelial, endothelial or fibroblastic cells to secrete IL-6, IL-8 and G-CSF. IL17 also increases the expression of ICAM-1 in fibroblasts.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.