Please login to your online account to display your discounted pricing

Gibco™ IL17 Recombinant Mouse Protein DFS Item


Recombinant IL17 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPMC0171

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PMC0171

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
Source Mouse
Protein Family Cytokines & Receptors
Gene ID (Entrez) 16171
Quantity 100μg
For Use With (Application) Cell Culture; Mammalian Cell Culture
Gene Alias IL-17
Purification Method Sequential Chromatography
Validated Application Cell Culture
Purity 95%
Structural Form Recombinant/Ligand
Protein Subtype Interleukins
Without Additives Carrier Protein
Storage Requirements Store at 2° to 8°C.
Molecular Weight 15kDa
Endotoxin Concentration <0.1ng/μg
Expression System E. coli

IL17 stimulates epithelial, endothelial or fibroblastic cells to secrete IL-6, IL-8 and G-CSF. IL17 also increases the expression of ICAM-1 in fibroblasts.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.