missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ IL13 Recombinant Mouse Protein DFS Item

Recombinant Mouse Interleukin 13 (IL13) is a bioactive protein intended for use in cell culture applications

$334.00 - $6,810.00

Specifications

Purity 98%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-13
View More Specs

Disclaimers:

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PMC0131 Gibco™
LSPMC0131
100μg Each for $1,418.00
PMC0133 Gibco™
LSPMC0133
1mg Each for $6,810.00
PMC0134 Gibco™
LSPMC0134
10μg Each for $334.00
PMC0135 Gibco™
LSPMC0135
25μg Each for $840.00
Specifications

Description

IL13 down-regulates macrophage produced cytokines such as TNF-α, IL-1, IL-6, IL-8, and IL-12 in response to LPS challenge or IFN-γ. IL13 induces monocyte differentiation and also induces B cell differentiation and proliferation.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human TF-1 cells

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

98%
<0.1ng/μg
IL-13
Sequential Chromatography
Lyophilized
16163