Please login to your online account to display your discounted pricing

Gibco™ IL12 Recombinant Human Protein DFS Item

Recombinant IL12 is a bioactive protein intended for use in cell culture applications

$566.00 - $566.00

Specifications

Purity 95%
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-12
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

PHC1123

DFS Item
 
gibco™
LSPHC1123
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PHC1124

DFS Item
 
gibco™
LSPHC1124
10μg Each for $566.00
Description & Specifications

Specifications

Purity 95%
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias IL-12

IL12 induces the secretion of IFN-γ, IL-2 and TNF-α and stimulates the proliferation of PHA activated lymphocytes.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.