Please login to your online account to display your discounted pricing

Gibco™ IL12 Recombinant Human Protein DFS Item


Recombinant IL12 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC1123

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC1123

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Molecular Weight 75kDa
Purity 95%
Quantity 1mg
Source Human
Storage Requirements Store at -5° to -30°C.
Without Additives Carrier Protein
Structural Form Recombinant/Ligand
Protein Subtype Interleukins
Endotoxin Concentration <0.1ng/μg
Gene Alias IL-12
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System Mammalian
Protein Length 503 aa
For Use With (Application) Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
Format Lyophilized

IL12 induces the secretion of IFN-γ, IL-2 and TNF-α and stimulates the proliferation of PHA activated lymphocytes.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.