Please login to your online account to display your discounted pricing

Gibco™ IL10 Recombinant Human Protein DFS Item


Recombinant IL10 is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPHC0103

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PHC0103

  • / Each
 

Description & Specifications

Specifications

Structural Form Recombinant/Ligand
Expression System E. coli
Molecular Weight 18.6kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 1mg
Source Human
Storage Requirements Store at 2° to 8°C.
Without Additives Carrier Protein
Protein Subtype Interleukins
Endotoxin Concentration <0.1ng/μg
Gene Alias IL-10
Validated Application Cell Culture
Protein Family Cytokines & Receptors
For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Angiogenesis; Cell Proliferation
Format Lyophilized

IL10 inhibits the synthesis of a number of cytokines, such as TNF-α, IL-1, and IL-6 from macrophages and IL-2, IFN-γ, and TNF-β from Th1 cells.

Approved for shipment on Wet or Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.