missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ IFN-γ Recombinant Rat Protein DFS Item

Recombinant IFN-γ is a bioactive protein intended for use in cell culture applications

$223.00 - $1,516.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Liquid
Gene Alias IFN-γ
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRC4033 Gibco™
LSPRC4033
1mg Each for $1,516.00
PRC4034 Gibco™
LSPRC4034
10μg Each for $223.00
Specifications

Description

Recombinant rat IFN-γ is intended for use in cell culture applications. IFN-γ is involved in many biological processes such as inducing the secretion of TNF-α and stimulating the release of reactive oxygen species from macrophages. It is also involved with bone growth and inhibits bone reesorption. It is anti-viral and anti-parasitic and inhibits the proliferation of cells.

Shipping Condition: Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

95%
<0.1ng/μg
IFN-γ
Sequential Chromatography
Liquid
25712