Please login to your online account to display your discounted pricing

Gibco™ IFN-γ Recombinant Rat Protein DFS Item


Recombinant IFN-γ is a bioactive protein intended for use in cell culture applications

Manufacturer: gibco™  LSPRC4034

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. PRC4034

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 16.9kDa
Purification Method Sequential Chromatography
Purity 95%
Quantity 10μg
Source Rat
Storage Requirements Store at -5° to -30°C.
Without Additives Carrier Protein
Structural Form Mutant/Recombinant/Ligand
Protein Subtype Interferons
Endotoxin Concentration <0.1ng/μg
Gene Alias IFN-γ
Validated Application Cell Culture
Protein Family Cytokines & Receptors
Expression System E. coli
For Use With (Application) Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Cell Signaling
Format Liquid
Gene ID (Entrez) 25712

Recombinant rat IFN-γ is intended for use in cell culture applications. IFN-γ is involved in many biological processes such as inducing the secretion of TNF-α and stimulating the release of reactive oxygen species from macrophages. It is also involved with bone growth and inhibits bone reesorption. It is anti-viral and anti-parasitic and inhibits the proliferation of cells.

Shipping Condition: Dry Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.