missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Mouse IFN-gamma Recombinant Protein
Recombinant Mouse Interferon-γ (IFN-γ) is a bioactive protein intended for use in cell culture applications
Supplier: Gibco™ PMC4033
Description
Mouse IFN-gamma Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IFN-γ is involved in many biological processes such as inducing the secretion of TNF-α and stimulating the release of reactive oxygen species from macrophages. It is also involved with bone growth and inhibits bone resorption. It is anti-viral and anti-parasitic and inhibits the proliferation of cells.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined in a dose response cytostasis assay of the mouse cell line WeHi279, as measured by the reduction of MTS
Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
∼15 kDa | |
1mg | |
Mouse | |
< 0.1 ng/ml | |
Dry Ice | |
Cytokines & Receptors | |
Liquid | |
Immunology, Virology, Oncology, Signal Transduction | |
95 % |
15978 | |
Sequential Chromatography | |
IFN-γ | |
Store in ultra-cold freezer (-68 to -85°C). | |
1 mg | |
E. coli | |
Recombinant, Ligand, Mutant | |
Interferons | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.